missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Bromo-3,5-dinitrobenzoic acid, 98%

Brand:  Alfa Aesar™ H64745.06

Additional Details : CAS Number : 116529-60-3 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15482059

  • 625.72 DKK / 5g


Estimated Shipment
26-10-2020
Add to basket

Specifications

Chemical Identifiers

116529-60-3
291.013
BBKJKUFARRJPRP-UHFFFAOYSA-N
3769937
C7H3BrN2O6
MFCD00458630
2-bromo-3,5-dinitrobenzoic acid, acmc-1bobs, 2-bromo-3,5-dinitrobenzoicacid, benzoic acid,2-bromo-3,5-dinitro
C1=C(C=C(C(=C1[N+](=O)[O-])Br)C(=O)O)[N+](=O)[O-]

Specifications

2-Bromo-3,5-dinitrobenzoic acid
White
5g
C7H3BrN2O6
2146483
BBKJKUFARRJPRP-UHFFFAOYSA-N
291.013
291.01
98%
Light sensitive
213°C to 216°C
116529-60-3
MFCD00458630
2-bromo-3,5-dinitrobenzoic acid, acmc-1bobs, 2-bromo-3,5-dinitrobenzoicacid, benzoic acid,2-bromo-3,5-dinitro
C1=C(C=C(C(=C1[N+](=O)[O-])Br)C(=O)O)[N+](=O)[O-]
3769937
Solid
Videos
SDS Documents