missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-3,5-dichloropyridine, 98%, Thermo Scientific™

Catalog Number 15487638
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
14482-51-0
C5H2BrCl2N
226.88
MFCD00233990
QCKPJWDIDCGQRB-UHFFFAOYSA-N
3,5-dichloro-2-bromopyridine, 2-bromo-3,5-dichloro-pyridine, pyridine, 2-bromo-3,5-dichloro, pubchem10223, acmc-1bqk7, 2-bromo-3,5-dichloropyridin, ksc494o1r, tpc-py058, 2-bromo-3,5 dichloro pyridine
817090
2-bromo-3,5-dichloropyridine
ClC1=CN=C(Br)C(Cl)=C1
This item is not returnable. View return policy
14482-51-0
C5H2BrCl2N
226.88
MFCD00233990
QCKPJWDIDCGQRB-UHFFFAOYSA-N
3,5-dichloro-2-bromopyridine, 2-bromo-3,5-dichloro-pyridine, pyridine, 2-bromo-3,5-dichloro, pubchem10223, acmc-1bqk7, 2-bromo-3,5-dichloropyridin, ksc494o1r, tpc-py058, 2-bromo-3,5 dichloro pyridine
817090
2-bromo-3,5-dichloropyridine
ClC1=CN=C(Br)C(Cl)=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Color Brown
Quantity 25 g
UN Number UN2811
Formula Weight 226.89
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 2-Bromo-3,5-dichloropyridine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'