missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-2-methylpropionic acid, 98%, Thermo Scientific Chemicals

Catalog Number p-3757085
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
2052-01-9
C4H7BrO2
167.01
MFCD00004168
XXSPGBOGLXKMDU-UHFFFAOYSA-N
α-Bromoisobutyric acid
2-bromo-2-methylpropanoic acid
CC(C)(Br)C(O)=O
This item is not returnable. View return policy
2052-01-9
C4H7BrO2
167.01
MFCD00004168
XXSPGBOGLXKMDU-UHFFFAOYSA-N
α-Bromoisobutyric acid
2-bromo-2-methylpropanoic acid
CC(C)(Br)C(O)=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 41°C to 46°C
Color Colorless to Yellow
Boiling Point 198°C to 200°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula BrC(CH3)2CO2H
Quantity 5 g
Merck Index 15, 1431
Solubility Information Solubility in water: slightly soluble
Formula Weight 167.01
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Bromo-2-methylpropionic acid
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'