Få mere at vide
2-Bromo-2-chloroacetonitrile (~90%), TRC
Artikelnummer. 30010195
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30010195 100 mg
30007874 50 mg
17922025 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30010195 Leverandør Toronto Research Chemicals Leverandørnr. TRCB685483100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 83463-62-1
Molekylær formel C2 H Br Cl N
Molekylvægt (g/mol) 154.39
Synonym Acetonitrile, bromochloro- (9CI), 2-Bromo-2-chloroacetonitrile, Bromochloroacetonitrile
IUPAC navn 2-bromo-2-chloroacetonitrile
SMIL ClC(Br)C#N
Tekniske data
Kemisk navn eller materiale Bromochloroacetonitrile
Mængde 100 mg
InChI formel InChI=1S/C2HBrClN/c3-2(4)1-5/h2H
Formel vægt 152.8981