missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Bromo-2',4'-dimethoxyacetophenone, 98%

Brand:  Alfa Aesar™ B22300.14

Additional Details : CAS Number : 60965-26-6 Weight : 0.02500kg Transport : UN number : 3261 Chem class : 8 Pack group : III

Quantity 25g

 View more versions of this product

Product Code. 11495976

  • 10469.00 DKK / 25g

Estimated Shipment
18-05-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

60965-26-6
259.099
PKVBZABQCCQHLD-UHFFFAOYSA-N
98683
COC1=CC(=C(C=C1)C(=O)CBr)OC
C10H11BrO3
MFCD00000197
2-bromo-1-2,4-dimethoxyphenyl ethanone, 2-bromo-2',4'-dimethoxyacetophenone, 2,4-dimethoxyphenacyl bromide, 2-bromo-1-2',4'-dimethoxyphenyl ethanone, 2-bromo-1-2,4-dimethoxyphenyl ethan-1-one, ethanone, 2-bromo-1-2,4-dimethoxyphenyl, 2-bromo-2',4'-dimethoxyacetopheneone, 1-2,4-dimethoxyphenyl-2-bromoethan-1-one, pubchem13433, acmc-1ba4l
2-bromo-1-(2,4-dimethoxyphenyl)ethanone

Specifications

2-Bromo-2',4'-dimethoxyacetophenone
25g
C10H11BrO3
UN3261
PKVBZABQCCQHLD-UHFFFAOYSA-N
2-bromo-1-(2,4-dimethoxyphenyl)ethanone
98683
98%
102°C to 105°C
60965-26-6
MFCD00000197
2-bromo-1-2,4-dimethoxyphenyl ethanone, 2-bromo-2',4'-dimethoxyacetophenone, 2,4-dimethoxyphenacyl bromide, 2-bromo-1-2',4'-dimethoxyphenyl ethanone, 2-bromo-1-2,4-dimethoxyphenyl ethan-1-one, ethanone, 2-bromo-1-2,4-dimethoxyphenyl, 2-bromo-2',4'-dimethoxyacetopheneone, 1-2,4-dimethoxyphenyl-2-bromoethan-1-one, pubchem13433, acmc-1ba4l
COC1=CC(=C(C=C1)C(=O)CBr)OC
259.099
259.1
Videos
SDS Documents