Få mere at vide
2-Bromo 17-Beta-estradiol, TRC
Artikelnummer. 30337969
Skift visning
Mængde:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30337969 500 mg
30347589 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30337969 Leverandør Toronto Research Chemicals Leverandørnr. TRCB684970500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Bromo 17beta-Estradiol
CAS 15833-07-5
Renhedsgrad noter HPLC
Molekylær formel C18H23BrO2
Mængde 500 mg
Synonym 2-Bromo-estradiol, 2-Bromo-1,3,5(10)-estratriene-3,17β-diol, 2-Bromo-β-estradiol, 2-Bromoestradiol, NSC 105460
InChI formel InChI=1S/C18H23BrO2/c1-18-7-6-11-12(14(18)4-5-17(18)21)3-2-10-8-16(20)15(19)9-13(10)11/h8-9,11-12,14,17,20-21H,2-7H2,1H3/t11-,12+,14-,17-,18-/m0/s1
SMIL C[C@]12CC[C@H]3[C@H]([C@@H]1CC[C@@H]2O)CCC4=CC(=C(C=C34)Br)O
IUPAC navn (8R,9S,13S,14S,17S)-2-bromo-13-methyl-6,7,8,9,11,12,14,15,16,17-decahydrocyclopenta[a]phenanthrene-3,17-diol
Molekylvægt (g/mol) 351.28
Formel vægt 350.0881
Procent renhed >95