Få mere at vide
2-Bromo-1,3-bis(tert-butyldimethylsilyloxy)propane-d4, TRC
Artikelnummer. 30336805
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30336805 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30336805 Leverandør Toronto Research Chemicals Leverandørnr. TRCB68186210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Bromo-1,3-bis(tert-butyldimethylsilyloxy)propane-d4
CAS 1794816-65-1
Molekylær formel C15H31D4BrO2Si2
Mængde 10 mg
InChI formel InChI=1S/C15H35BrO2Si2/c1-14(2,3)19(7,8)17-11-13(16)12-18-20(9,10)15(4,5)6/h13H,11-12H2,1-10H3/i11D2,12D2
SMIL [2H]C([2H])(O[Si](C)(C)C(C)(C)C)C(Br)C([2H])([2H])O[Si](C)(C)C(C)(C)C
IUPAC navn [2-bromo-3-[tert-butyl(dimethyl)silyl]oxy-1,1,3,3-tetradeuteriopropoxy]-tert-butyl-dimethylsilane
Molekylvægt (g/mol) 387.54
Formel vægt 386.161