missing translation for 'onlineSavingsMsg'
Få mere at vide

2-Bromo-1,3,4-thiadiazole, TRC

Artikelnummer. 30338649
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
250 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30338649 250 mg
30331878 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30338649 Leverandør Toronto Research Chemicals Leverandørnr. TRCB688338250MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale 2-Bromo-1,3,4-thiadiazole
CAS 61929-24-6
Molekylær formel C2HBrN2S
Mængde 250 mg
InChI formel InChI=1S/C2HBrN2S/c3-2-5-4-1-6-2/h1H
SMIL Brc1nncs1
IUPAC navn 2-bromo-1,3,4-thiadiazole
Molekylvægt (g/mol) 165.01
Formel vægt 163.9044
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.