Learn More
Alfa Aesar™ 2-(Boc-amino)benzoic acid, 98%
Brand:  Alfa Aesar™ H61780.06
Additional Details : CAS Number : 68790-38-5 Weight : 0.00500kg
Quantity 5g
 View more versions of this product
Product Code. 15408358
  • 3110.84 DKK / 5g
Estimated Shipment date
from Supplier 31-08-2018
Add to basket
Specifications
Chemical Identifiers
68790-38-5
237.255
BYGHHEDJDSLEKK-UHFFFAOYSA-N
4087778
C12H15NO4
MFCD00151862
Boc-2-Abz-OH; N-Boc-anthranilic acid
Specifications
2-(Boc-amino)benzoic acid
98%
153°C to 156°C (decomposition)
5g
Boc-2-Abz-OH; N-Boc-anthranilic acid
CC(C)(C)OC(=O)NC1=CC=CC=C1C(=O)O
237.255
237.25
68790-38-5
C12H15NO4
MFCD00151862
2848891
BYGHHEDJDSLEKK-UHFFFAOYSA-N
2-[(2-methylpropan-2-yl)oxycarbonylamino]benzoic acid
4087778
Solid