missing translation for 'onlineSavingsMsg'
Learn More

2-[Bis(3,5-di-tert-butyl-4-methoxyphenyl)phosphino]benzaldehyde, 97%, Thermo Scientific Chemicals

Catalog Number p-7055406
Click to view available options
Quantity:
100 mg
500 mg
This item is not returnable. View return policy
1202865-21-1
C37H51O3P
574.786
MFCD15143663
PAWZHFURVJBYGV-UHFFFAOYSA-N
2-bis 3,5-di-tert-butyl-4-methoxyphenyl phosphino benzaldehyde, 2-bis 3,5-di-t-butyl-4-methoxyphenyl phosphino benzaldehyde, 2-bis 3,5-di-tert-butyl-4-methoxyphenyl phosphanyl benzaldehyde, acmc-20ao6l, 2-bis 3,5-di-t-butyl-4-methoxyphenyl phosphino benzaldehyde, min
71310909
2-bis(3,5-ditert-butyl-4-methoxyphenyl)phosphanylbenzaldehyde
CC(C)(C)C1=CC(=CC(=C1OC)C(C)(C)C)P(C2=CC=CC=C2C=O)C3=CC(=C(C(=C3)C(C)(C)C)OC)C(C)(C)C
This item is not returnable. View return policy
1202865-21-1
C37H51O3P
574.786
MFCD15143663
PAWZHFURVJBYGV-UHFFFAOYSA-N
2-bis 3,5-di-tert-butyl-4-methoxyphenyl phosphino benzaldehyde, 2-bis 3,5-di-t-butyl-4-methoxyphenyl phosphino benzaldehyde, 2-bis 3,5-di-tert-butyl-4-methoxyphenyl phosphanyl benzaldehyde, acmc-20ao6l, 2-bis 3,5-di-t-butyl-4-methoxyphenyl phosphino benzaldehyde, min
71310909
2-bis(3,5-ditert-butyl-4-methoxyphenyl)phosphanylbenzaldehyde
CC(C)(C)C1=CC(=CC(=C1OC)C(C)(C)C)P(C2=CC=CC=C2C=O)C3=CC(=C(C(=C3)C(C)(C)C)OC)C(C)(C)C

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 120°C to 125°C
Color Yellow
Quantity 100 mg
Sensitivity Air Sensitive
Formula Weight 574.77
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 2-[Bis(3,5-di-tert-butyl-4-methoxyphenyl)phosphino]benzaldehyde

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'