missing translation for 'onlineSavingsMsg'
Få mere at vide

2-Benzylamino-2-methyl-1-propanol, TRC

Artikelnummer. 30337072
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30337072 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30337072 Supplier Toronto Research Chemicals Supplier No. TRCB224949250MG
 Begrænset vare
Forventet forsendelse: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Højrenheds organiske molekyler og analytiske standarder, strategisk leveret verden over for at muliggøre innovation og kommerciel succes.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 2-Benzylamino-2-methyl-1-propanol
CAS 10250-27-8
Molekylær formel C11 H17 N O
Mængde 250 mg
Synonym 1-Propanol, 2-methyl-2-[(phenylmethyl)amino]-, 1-Propanol, 2-(benzylamino)-2-methyl- (7CI,8CI), 2-Methyl-2-[(phenylmethyl)amino]-1-propanol, (Benzyl)(2-hydroxy-1,1-dimethylethyl)amine, 2-(Benzylamino)-2-methyl-1-propanol, 2-(Benzylamino)-2-methyl-propanol
InChI formel InChI=1S/C11H17NO/c1-11(2,9-13)12-8-10-6-4-3-5-7-10/h3-7,12-13H,8-9H2,1-2H3
SMIL CC(C)(CO)NCc1ccccc1
IUPAC navn 2-(benzylamino)-2-methylpropan-1-ol
Molekylvægt (g/mol) 179.26
Formel vægt 179.131
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.