Få mere at vide
2-(Benzyl(1-(4-methoxyphenyl)propan-2-yl)amino)-1-(4-(benzyloxy)-3-nitrophenyl)ethanone, TRC
Artikelnummer. 30347880
Skift visning
Mængde:
50 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30347880 250 mg
30340124 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30347880 Leverandør Toronto Research Chemicals Leverandørnr. TRCB285565250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-(Benzyl(1-(4-methoxyphenyl)propan-2-yl)amino)-1-(4-(benzyloxy)-3-nitrophenyl)ethanone
CAS 43229-66-9
Molekylær formel C32H32N2O5
Mængde 250 mg
Synonym 2-[[2-(4-Methoxyphenyl)-1-methylethyl](phenylmethyl)amino]-1-[3-nitro-4-(phenylmethoxy)phenyl]ethanone
InChI formel InChI=1S/C32H32N2O5/c1-24(19-25-13-16-29(38-2)17-14-25)33(21-26-9-5-3-6-10-26)22-31(35)28-15-18-32(30(20-28)34(36)37)39-23-27-11-7-4-8-12-27/h3-18,20,24H,19,21-23H2,1-2H3
SMIL COc1ccc(CC(C)N(CC(=O)c2ccc(OCc3ccccc3)c(c2)[N+](=O)[O-])Cc4ccccc4)cc1
IUPAC navn 2-[benzyl-[1-(4-methoxyphenyl)propan-2-yl]amino]-1-(3-nitro-4-phenylmethoxyphenyl)ethanone
Molekylvægt (g/mol) 524.61
Formel vægt 524.2311