Learn More
2-Benzoylpyrrole, TRC
Specifications
Specifications
| Chemical Name or Material | 2-Benzoylpyrrole |
| CAS | 7697-46-3 |
| Molecular Formula | C11 H9 N O |
| Quantity | 1 g |
| Synonym | Pyrrol-2-yl Phenyl Ketone, Phenyl(1H-pyrrol-2-yl)methanone, 2-Benzoyl-1H-pyrrole |
| InChI Formula | InChI=1S/C11H9NO/c13-11(10-7-4-8-12-10)9-5-2-1-3-6-9/h1-8,12H |
| SMILES | O=C(c1ccccc1)c2ccc[nH]2 |
| IUPAC Name | phenyl(1H-pyrrol-2-yl)methanone |
| Molecular Weight (g/mol) | 171.2 |
| Formula Weight | 171.0684 |