Få mere at vide
2-Benzoylbenzoic acid-methyl ester, Dr. Ehrenstorfer
Tekniske data
Tekniske data
| Kemisk navn eller materiale | 2-Benzoylbenzoic acid methyl ester |
| CAS | 606-28-0 |
| Analyt- eller komponentnavne | 2-Benzoylbenzoic acid methyl ester |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C15 H12 O3 |
| Certificeringer/compliance | ISO/IEC 17025 |
| Synonym | Benzoic acid, o-benzoyl-, methyl ester (6CI,7CI,8CI), 2-Benzoylbenzoic acid methyl ester, Daitocure OB, Genocure MBB, Initiator 1056, Methyl 2-benzoylbenzoate, Methyl o-benzoylbenzoate, NSC 3797, OBM 100, OMBB, Omnirad OMBB, Runtecure 1056, SB-PI 711, Speedcure MBB, o-(Methoxycarbonyl)benzophenone |
| InChI formel | InChI=1S/C15H12O3/c1-18-15(17)13-10-6-5-9-12(13)14(16)11-7-3-2-4-8-11/h2-10H,1H3 |
| Løsningstype | Neat |
| SMIL | COC(=O)c1ccccc1C(=O)c2ccccc2 |
| Vis mere |