missing translation for 'onlineSavingsMsg'
Learn More

2-Benzamido-1,4-naphthoquinone, TRC

Product Code. 30341902
Change view
Click to view available options
Mængde:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30341902 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30341902 Supplier Toronto Research Chemicals Supplier No. TRCB18499010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 2-Benzamido-1,4-naphthoquinone
CAS 65240-86-0
Molekylær formel C17H11NO3
Mængde 10 mg
Synonym N-(1,4-Dihydro-1,4-dioxo-2-naphthalenyl)benzamide, N-(1,4-Dioxo-1,4-dihydronaphthalen-2-yl)benzamide, NSC 73233, PPM 18
InChI formel InChI=1S/C17H11NO3/c19-15-10-14(16(20)13-9-5-4-8-12(13)15)18-17(21)11-6-2-1-3-7-11/h1-10H,(H,18,21)
SMIL O=C(NC1=CC(=O)c2ccccc2C1=O)c3ccccc3
IUPAC navn N-(1,4-dioxonaphthalen-2-yl)benzamide
Molekylvægt (g/mol) 277.27
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.