missing translation for 'onlineSavingsMsg'
Få mere at vide

2-Azido-2-deoxy-D-glucopyranose 1,3,4,6-Tetraacetate, TRC

Artikelnummer. 30331486
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
250 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30331486 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30331486 Leverandør Toronto Research Chemicals Leverandørnr. TRCA824855250G

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 2-Azido-2-deoxy-D-glucopyranose 1,3,4,6-Tetraacetate
CAS 171032-74-9
Molekylær formel C14 H19 N3 O9
Mængde 250 g
Synonym Acetyl 2-Azido-2-deoxy-3,4,6-tri-O-acetyl-D-glucopyranoside
InChI formel InChI=1S/C14H19N3O9/c1-6(18)22-5-10-12(23-7(2)19)13(24-8(3)20)11(16-17-15)14(26-10)25-9(4)21/h10-14H,5H2,1-4H3/t10-,11-,12-,13-,14?/m1/s1
SMIL CC(=O)OC[C@H]1OC(OC(=O)C)[C@H](N=[N+]=[N-])[C@@H](OC(=O)C)[C@@H]1OC(=O)C
IUPAC navn [(2R,3S,4R,5R)-3,4,6-triacetyloxy-5-azidooxan-2-yl]methyl acetate
Molekylvægt (g/mol) 373.32
Formel vægt 373.1121
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.