Få mere at vide
2-Arsanilic Acid, TRC
Artikelnummer. 30344127
Skift visning
Mængde:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30344127 1 g
30334928 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30344127 Leverandør Toronto Research Chemicals Leverandørnr. TRCA7741001G
Denne vare kræver en prisopdatering, inden den kan bestilles.
Anmod om en pris på denne vare her.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Arsanilic Acid
CAS 2045-00-3
Molekylær formel C6 H8 As N O3
Mængde 1 g
Synonym (2-Aminophenyl)arsonic Acid, (o-Aminophenyl)arsonic Acid, 2-Aminobenzenearsonic Acid, o-Arsanilic Acid, NSC 12611, o-Aminobenzenearsonic Acid
InChI formel InChI=1S/C6H8AsNO3/c8-6-4-2-1-3-5(6)7(9,10)11/h1-4H,8H2,(H2,9,10,11)
SMIL Nc1ccccc1[As](=O)(O)O
IUPAC navn (2-aminophenyl)arsonic acid
Molekylvægt (g/mol) 217.05
Formel vægt 216.972