Få mere at vide
2-Arachidonoyl-sn-glycero-3-phosphocholine (>90%), TRC
Artikelnummer. 30132901
Skift visning
Mængde:
2.5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30132901 2.5 mg
30137043 25 mg
30011371 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30132901 Leverandør Toronto Research Chemicals Leverandørnr. TRCA7650902.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 67341-29-1
Molekylær formel C28H50NO7P
Molekylvægt (g/mol) 543.68
Synonym (7R,13Z,16Z,19Z,22Z)-4-Hydroxy-7-(hydroxymethyl)-N,N,N-trimethyl-9-oxo-3,5,8-trioxa-4-phosphaoctacosa-13,16,19,22-tetraen-1-aminium 4-Oxide Inner Salt, [R-(all-Z)]-4-Hydroxy-7-(hydroxymethyl)-N,N,N-trimethyl-9-oxo-3,5,8-trioxa-4-phosphaoctacosa-13,16,19,22-tetraen-1-aminium 4-Oxide Inner Salt
IUPAC navn (R)-3-hydroxy-2-(((5Z,8Z,11Z,14Z)-icosa-5,8,11,14-tetraenoyl)oxy)propyl (2-(trimethylammonio)ethyl) phosphate
SMIL O=C(CCC/C=C\C/C=C\C/C=C\C/C=C\CCCCC)O[C@@H](COP([O-])(OCC[N+](C)(C)C)=O)CO
Tekniske data
Kemisk navn eller materiale 2-Arachidonoyl-sn-glycero-3-phosphocholine
Analyt- eller komponentnavne 2-Arachidonoyl-sn-glycero-3-phosphocholine
Forsendelsestilstand Dry ice
InChI formel InChI=1S/C28H50NO7P/c1-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22-28(31)36-27(25-30)26-35-37(32,33)34-24-23-29(2,3)4/h9-10,12-13,15-16,18-19,27,30H,5-8,11,14,17,20-26H2,1-4H3/b10-9-,13-12-,16-15-,19-18-/t27-/m1/s1
Løsningstype Neat
Industry Type Forensics
Mængde 2.5 mg
Formel vægt 543.33