Få mere at vide
2-Aminopyridine-d6, TRC
Artikelnummer. 30308901
Skift visning
Mængde:
2 mg
Artikelnummer. Mængde
30308901 2 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30308901 Leverandør Toronto Research Chemicals Leverandørnr. TRCA6299732MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Aminopyridine-d6
CAS 203784-57-0
Molekylær formel C5 D6 N2
Mængde 2 mg
Synonym 2-Pyridin-3,4,5,6-d4-amine-d2 (9CI, ACI), 2-Aminopyridine-d6, 2-Aminopyridine-D6, 2-Pyridyl-d4-amine-d2, 2-Pyridinamine-d6
InChI formel InChI=1S/C5H6N2/c6-5-3-1-2-4-7-5/h1-4H,(H2,6,7)/i1D,2D,3D,4D/hD2
SMIL [2H]N([2H])c1nc([2H])c([2H])c([2H])c1[2H]
IUPAC navn N,N,3,4,5,6-hexadeuteriopyridin-2-amine
Molekylvægt (g/mol) 100.151
Formel vægt 100.091