Få mere at vide
2-Aminonaphthalene-d7, CDN
Artikelnummer. 30319549
Skift visning
Mængde:
0.05 g
0.1 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30319549 0.05 g -
30318565 0.1 g 0.1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30319549 Leverandør CDN Leverandørnr. CDND01910.05G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Naphthylamine-d7
CAS 93951-94-1
Molekylær formel C10 2H7 H2 N
Mængde 0.05 g
Synonym 2-Naphthalen-1,3,4,5,6,7,8-d7-amine, 2-Aminonaphthalene D7, 2-Naphthylamine-d7
InChI formel InChI=1 S/C10H9N/c11-10-6-5-8-3-1-2-4-9(8)7-10/h1-7 H,11H2/i1D,2 D,3 D,4 D,5 D,6 D,7 D
SMIL [2 H]c1c([2 H])c([2 H])c2c([2 H])c(N)c([2 H])c([2 H])c2c1[2 H]
IUPAC navn 1,3,4,5,6,7,8-heptadeuterionaphthalen-2-amine
Molekylvægt (g/mol) 150.23
Formel vægt 150.1174 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity