missing translation for 'onlineSavingsMsg'
Learn More

2-Aminoisobutyric acid methyl ester hydrochloride, 99%, Thermo Scientific Chemicals

Catalog Number p-7029116
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
15028-41-8
C5H12ClNO2
153.606
MFCD00214247
NVWZNEDLYYLQJC-UHFFFAOYSA-N
methyl 2-amino-2-methylpropanoate hydrochloride, methyl 2-aminoisobutyrate hydrochloride, h-aib-ome.hcl, alpha-aminoisobutyric acid methyl ester hydrochloride, 2-aminoisobutyric acid methyl ester hydrochloride, methyl alpha-aminoisobutyrate hydrochloride, methyl 2-methylalaninate hydrochloride, alanine, 2-methyl-, methyl ester, hydrochloride, aib-ome hydrochloride, pubchem16472
13258034
methyl 2-amino-2-methylpropanoate;hydrochloride
CC(C)(C(=O)OC)N.Cl
This item is not returnable. View return policy
15028-41-8
C5H12ClNO2
153.606
MFCD00214247
NVWZNEDLYYLQJC-UHFFFAOYSA-N
methyl 2-amino-2-methylpropanoate hydrochloride, methyl 2-aminoisobutyrate hydrochloride, h-aib-ome.hcl, alpha-aminoisobutyric acid methyl ester hydrochloride, 2-aminoisobutyric acid methyl ester hydrochloride, methyl alpha-aminoisobutyrate hydrochloride, methyl 2-methylalaninate hydrochloride, alanine, 2-methyl-, methyl ester, hydrochloride, aib-ome hydrochloride, pubchem16472
13258034
methyl 2-amino-2-methylpropanoate;hydrochloride
CC(C)(C(=O)OC)N.Cl

Methyl α-aminoisobutyrate is used as a specific substrate of amino acid transport system and in studies on amino acid transport systems.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl α-aminoisobutyrate is used as a specific substrate of amino acid transport system and in studies on amino acid transport systems.

Solubility
Slightly soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY
Melting Point ∽185°C (decomposition)
Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 153.61
Percent Purity 99%
Chemical Name or Material 2-Aminoisobutyric acid methyl ester hydrochloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'