missing translation for 'onlineSavingsMsg'
Få mere at vide

2-Aminoindole Hydrochloride, TRC

Artikelnummer. 30312932
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30312932 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30312932 Supplier Toronto Research Chemicals Supplier No. TRCA611707500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 2-Aminoindole, Hydrochloride
CAS 27878-37-1
Molekylær formel C8 H8 N2 . Cl H
Mængde 500 mg
InChI formel InChI=1S/C8H8N2.ClH/c9-8-5-6-3-1-2-4-7(6)10-8;/h1-5,10H,9H2;1H
SMIL Cl.Nc1cc2ccccc2[nH]1
IUPAC navn 1H-indol-2-amine;hydrochloride
Molekylvægt (g/mol) 168.62
Formel vægt 168.0454
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.