Få mere at vide
2-Aminofluorene-d11, TRC
Artikelnummer. 30326880
Skift visning
Mængde:
1 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30326880 1 mg
30310083 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30326880 Leverandør Toronto Research Chemicals Leverandørnr. TRCA6082971MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Aminofluorene-D11
CAS 347841-44-5
Molekylær formel C13 D11 N
Mængde 1 mg
Synonym 9H-Fluoren-1,3,4,5,6,7,8,9,9-d9-2-amine-d2 (9CI), 2-Aminofluorene-d11, 2-Aminofluorene-D11
InChI formel InChI=1S/C13H11N/c14-11-5-6-13-10(8-11)7-9-3-1-2-4-12(9)13/h1-6,8H,7,14H2/i1D,2D,3D,4D,5D,6D,7D2,8D/hD2
SMIL [2H]N([2H])c1c([2H])c([2H])c2c3c([2H])c([2H])c([2H])c([2H])c3C([2H])([2H])c2c1[2H]
IUPAC navn N,N,1,3,4,5,6,7,8,9,9-undecadeuteriofluoren-2-amine
Molekylvægt (g/mol) 192.3009
Formel vægt 192.1582