Få mere at vide
2-Aminofluorene-9-13C, TRC
Artikelnummer. 30320058
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30320058 1 mg
30312782 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30320058 Leverandør Toronto Research Chemicals Leverandørnr. TRCA6082991MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Aminofluorene-9-13C
CAS 335081-08-8
Molekylær formel 13C C12 H11 N
Mængde 1 mg
Synonym 9H-Fluoren-2-amine-9-13C (9CI), 2-Aminofluorene-9-13C, 2-Aminofluorene-9-13C1
InChI formel InChI=1S/C13H11N/c14-11-5-6-13-10(8-11)7-9-3-1-2-4-12(9)13/h1-6,8H,7,14H2/i7+1
SMIL Nc1ccc2c([13CH2]c3ccccc23)c1
IUPAC navn 9H-fluoren-2-amine
Molekylvægt (g/mol) 182.2258
Formel vægt 182.0925