Få mere at vide
(2-Aminoethyl)trimethylammonium Chloride Hydrochloride, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | (2-Aminoethyl)trimethylammonium Chloride Hydrochloride |
| CAS | 3399-67-5 |
| Molekylær formel | C5H15N2ClHCl |
| Mængde | 500 mg |
| Synonym | Ethanaminium, 2-amino-N,N,N-trimethyl-, chloride, hydrochloride (1:1:1) (ACI), (2-Aminoethyl)trimethylammonium chloride, hydrochloride (7CI), Ammonium, (2-aminoethyl)trimethyl-, chloride, hydrochloride (8CI), Ethanaminium, 2-amino-N,N,N-trimethyl-, chloride, hydrochloride (9CI), (2-Aminoethyl)trimethylazanium hydrochloride chloride, 1-Amino-2-(trimethylammonium)ethane chloride, 2-AETA, 2-Amino-N,N,N-trimethylethanaminium chloride hydrochloride, Cholamine chloride hydrochloride, (2-Aminoethyl)trimethylammonium chloride hydrochloride |
| InChI formel | InChI=1S/C5H15N2.2ClH/c1-7(2,3)5-4-6;;/h4-6H2,1-3H3;2*1H/q+1;;/p-1 |
| SMIL | Cl.[Cl-].C[N+](C)(C)CCN |
| IUPAC navn | 2-aminoethyl(trimethyl)azanium;chloride;hydrochloride |
| Molekylvægt (g/mol) | 175.1 |
| Formel vægt | 174.0691 |