missing translation for 'onlineSavingsMsg'
Få mere at vide

2-Aminoethyl Methanethiosulfonate Hydrochloride, TRC

Artikelnummer. 30309335
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30309335 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30309335 Leverandør Toronto Research Chemicals Leverandørnr. TRCA60915050MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 2-Aminoethyl Methanethiosulfonate, Hydrochloride
CAS 37597-96-9
Molekylær formel C3 H9 N O2 S2 . Cl H
Mængde 50 mg
InChI formel InChI=1S/C3H9NO2S2.ClH/c1-8(5,6)7-3-2-4;/h2-4H2,1H3;1H
SMIL Cl.CS(=O)(=O)SCCN
IUPAC navn 2-methylsulfonylsulfanylethanamine;hydrochloride
Molekylvægt (g/mol) 191.7
Formel vægt 190.9841
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.