Få mere at vide
2-Aminoethyl Methanethiosulfonate Hydrobromide, TRC
Artikelnummer. 30132521
Skift visning
Mængde:
250 mg
2.5 g
Pakningsstørrelse:
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30132521 2.5 g 2.5g
30008414 250 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30132521 Leverandør Toronto Research Chemicals Leverandørnr. TRCA6091002.5G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 16599-33-0
Molekylær formel C3 H9 N O2 S2 . Br H
Molekylvægt (g/mol) 236.15
IUPAC navn 2-methylsulfonylsulfanylethanamine;hydrobromide
SMIL Br.CS(=O)(=O)SCCN
Tekniske data
Kemisk navn eller materiale 2-Aminoethyl Methanethiosulfonate, Hydrobromide
Analyt- eller komponentnavne 2-Aminoethyl Methanethiosulfonate, Hydrobromide
Renhedsgrad noter HPLC
Forsendelsestilstand Room Temperature
InChI formel InChI=1S/C3H9NO2S2.BrH/c1-8(5,6)7-3-2-4;/h2-4H2,1H3;1H
Løsningstype Neat
Industry Type Pharmaceutical
Mængde 2.5 g
Formel vægt 234.9336
Procent renhed >95%