Learn More
2-Aminoethyl Diphenylborinate, TRC
Product Code. 30011621
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
Unit Size:
100mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30011621 100 mg 100mg
30010624 5 g 5g
30012118 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30011621 Supplier Toronto Research Chemicals Supplier No. TRCA609115100MG
 Restricted Item
Forventet forsendelse: 03-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 524-95-8
Molecular Formula C14H16BNO
Molecular Weight (g/mol) 225.09
Synonym Diphenylborinic Acid 2-Aminoethyl Ester, 2-Aminoethanol Diphenylborinate (Ester), (2-Aminoethyloxy)diphenylborane, 2-APB, 2-Aminoethoxydiphenyl Borate, 2-Aminoethoxydiphenylborane, 2-Aminoethyl Diphenylborinate, B-(2- Aminoethoxy)diphenylborane, NSC 17107, Naturstoffreagenz A
IUPAC Name 2-diphenylboranyloxyethanamine
SMILES NCCOB(c1ccccc1)c2ccccc2
Specifications
Quantity 100 mg
InChI Formula InChI=1S/C14H16BNO/c16-11-12-17-15(13-7-3-1-4-8-13)14-9-5-2-6-10-14/h1-10H,11-12,16H2
Formula Weight 225.1325
Chemical Name or Material 2-Aminoethyl Diphenylborinate