Learn More
2-Aminoethyl diphenylborinate, 98%, Thermo Scientific Chemicals
434.72 DKK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
5 g
25 g
Description
2-Aminoethyl diphenylborinate is used for the densitometric determination of flavonoids and TRP activator. It is also used to manipulate intracellular release of calcium ions and in preparation of apoptotic inducers. It serves as a reactant for preparation of apoptotic inducers and in membrane-permeable SOCE (store operated calcuym entry) inhibitor.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Aminoethyl diphenylborinate is used for the densitometric determination of flavonoids and TRP activator. It is also used to manipulate intracellular release of calcium ions and in preparation of apoptotic inducers. It serves as a reactant for preparation of apoptotic inducers and in membrane-permeable SOCE (store operated calcuym entry) inhibitor.
Solubility
Soluble in ethanol and methanol.
Notes
Incompatible with strong oxidizing agents.
2-Aminoethyl diphenylborinate is used for the densitometric determination of flavonoids and TRP activator. It is also used to manipulate intracellular release of calcium ions and in preparation of apoptotic inducers. It serves as a reactant for preparation of apoptotic inducers and in membrane-permeable SOCE (store operated calcuym entry) inhibitor.
Solubility
Soluble in ethanol and methanol.
Notes
Incompatible with strong oxidizing agents.
Specifications
Melting Point | 187°C to 190°C |
Quantity | 5 g |
Beilstein | 2942273 |
Solubility Information | Soluble in ethanol and methanol. |
IUPAC Name | 2-aminoethyl diphenylborinate |
Formula Weight | 225.1 |
Percent Purity | 98% |
Chemical Name or Material | 2-Aminoethyl diphenylborinate |