missing translation for 'onlineSavingsMsg'
Få mere at vide

2-Aminoethyl 4-tert-Butyl-2,6-dimethyl-3-hydroxyphenylacetamide Hydrochloride, TRC

Artikelnummer. 30318310
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30318310 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.
Artikelnummer. 30318310 Leverandør Toronto Research Chemicals Leverandørnr. TRCA60889010MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale N-(2-Amino-ethyl)-2-[4-(1,1-dimethylethyl)-3-hydroxy-2,6-dimethylphenyl]acetamide Hydrochloride
CAS 1391053-50-1
Molekylær formel C16 H26 N2 O2 . Cl H
Mængde 10 mg
Synonym N-(2-Amino-ethyl)-2-[4-(1,1-dimethylethyl)-3-hydroxy-2,6-dimethylphenyl]acetamide Hydrochloride, Oxymetazoline Hydrochloride Imp. A (EP) as Hydrochloride, Benzeneacetamide, N-(2-aminoethyl)-4-(1,1-dimethylethyl)-3-hydroxy-2,6-dimethyl-, hydrochloride (1:1)
InChI formel InChI=1S/C16H26N2O2.ClH/c1-10-8-13(16(3,4)5)15(20)11(2)12(10)9-14(19)18-7-6-17;/h8,20H,6-7,9,17H2,1-5H3,(H,18,19);1H
SMIL Cl.Cc1cc(c(O)c(C)c1CC(=O)NCCN)C(C)(C)C
IUPAC navn N-(2-aminoethyl)-2-(4-tert-butyl-3-hydroxy-2,6-dimethylphenyl)acetamide;hydrochloride
Molekylvægt (g/mol) 314.85
Formel vægt 314.1761
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.