Få mere at vide
2-Aminobiphenyl-d9, TRC
Artikelnummer. 30322250
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30322250 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30322250 Leverandør Toronto Research Chemicals Leverandørnr. TRCA6017661MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Aminobiphenyl-d9
CAS 344298-97-1
Molekylær formel C12 D9 H2 N
Mængde 1 mg
Synonym 2-Aminobiphenyl-d9
InChI formel InChI=1S/C12H11N/c13-12-9-5-4-8-11(12)10-6-2-1-3-7-10/h1-9H,13H2/i1D,2D,3D,4D,5D,6D,7D,8D,9D
SMIL [2H]c1c([2H])c([2H])c(c([2H])c1[2H])c2c([2H])c([2H])c([2H])c([2H])c2N
IUPAC navn 2,3,4,5-tetradeuterio-6-(2,3,4,5,6-pentadeuteriophenyl)aniline
Molekylvægt (g/mol) 178.2779
Formel vægt 178.1456