Få mere at vide
2-Aminobiphenyl-d9, CDN
Artikelnummer. 30320799
Skift visning
Mængde:
0.05 g
0.1 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30320799 0.05 g -
30312926 0.1 g 0.1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30320799 Leverandør CDN Leverandørnr. CDND23310.05G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Aminobiphenyl-d9
CAS 344298-97-1
Molekylær formel C12 D9 H2 N
Mængde 0.05 g
Synonym [1,1'-Biphenyl-2',3,3',4,4',5,5',6,6'-d9]-2-amine (9 CI), 2-Aminobiphenyl-d9, 2-Aminobiphenyl-D9, 2-(Phenyl-d5)aniline-d4, 2-Biphenylamine-2',3,3',4,4',5,5',6,6'-d9
InChI formel InChI=1 S/C12H11N/c13-12-9-5-4-8-11(12)10-6-2-1-3-7-10/h1-9 H,13H2/i1D,2 D,3 D,4 D,5 D,6 D,7 D,8 D,9 D
SMIL [2 H]c1c([2 H])c([2 H])c(c([2 H])c1[2 H])c2c([2 H])c([2 H])c([2 H])c([2 H])c2N
IUPAC navn 2,3,4,5-tetradeuterio-6-(2,3,4,5,6-pentadeuteriophenyl)aniline
Molekylvægt (g/mol) 178.2779
Formel vægt 178.1456 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity