missing translation for 'onlineSavingsMsg'
Få mere at vide

2-Aminobenzoxazol, TRC

Artikelnummer. 30322453
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30322453 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Product Code. 30322453 missing translation for 'mfr' Toronto Research Chemicals missing translation for 'supplierNo' TRCA5915155G
 Begrænset vare
Forventet forsendelse: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Højrenheds organiske molekyler og analytiske standarder, strategisk leveret verden over for at muliggøre innovation og kommerciel succes.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 1,3-Benzoxazol-2-amine
CAS 4570-41-6
Molekylær formel C7 H6 N2 O
Mængde 5 g
Synonym 2-Benzoxazolamine, 1,3-Benzoxazol-2-amine
InChI formel InChI=1S/C7H6N2O/c8-7-9-5-3-1-2-4-6(5)10-7/h1-4H,(H2,8,9)
SMIL Nc1oc2ccccc2n1
IUPAC navn 1,3-benzoxazol-2-amin
Molekylvægt (g/mol) 134.14
Formel vægt 134.048
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.