missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Aminobenzophenone, 98%, Thermo Scientific Chemicals
208.67 DKK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
10 g
50 g
250 g
Description
It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and dyestuffs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and dyestuffs.
Solubility
Insoluble in water. Soluble in DMSO, Methanol.
Notes
Store at room temperature. Ensure good ventilation.
It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and dyestuffs.
Solubility
Insoluble in water. Soluble in DMSO, Methanol.
Notes
Store at room temperature. Ensure good ventilation.
Specifications
Melting Point | 105°C to 108°C |
Boiling Point | 280°C |
Flash Point | 240°C (464°F) |
Quantity | 10 g |
Beilstein | 743425 |
Solubility Information | Insoluble in water. Soluble in DMSO,Methanol. |
Formula Weight | 197.24 |
Percent Purity | 98% |
Chemical Name or Material | 2-Aminobenzophenone |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'