Få mere at vide
2-Amino-N-benzylbenzamide, TRC
Artikelnummer. 30319504
Skift visning
Mængde:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30319504 100 mg
30316522 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30319504 Leverandør Toronto Research Chemicals Leverandørnr. TRCA630243100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Amino-N-benzylbenzamide
CAS 5471-20-5
Molekylær formel C14H14N2O
Mængde 100 mg
Synonym 2-Amino-N-(phenylmethyl)benzamide, 2-(Benzylaminocarbonyl)aniline, Anthranilic Acid Benzylamide
InChI formel InChI=1S/C14H14N2O/c15-13-9-5-4-8-12(13)14(17)16-10-11-6-2-1-3-7-11/h1-9H,10,15H2,(H,16,17)
SMIL Nc1ccccc1C(=O)NCc2ccccc2
IUPAC navn 2-amino-N-benzylbenzamide
Molekylvægt (g/mol) 226.27
Formel vægt 226.1106