Få mere at vide
2-Amino-Alpha-(methoxyimino)-4-thiazoleacetic Acid-d3, TRC
Artikelnummer. 30313546
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30313546 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30313546 Leverandør Toronto Research Chemicals Leverandørnr. TRCA6119671MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Amino-alpha-(methoxyimino)-4-thiazoleacetic Acid-d3
CAS 1331655-54-9
Molekylær formel C6H4D3N3O3S
Mængde 1 mg
Synonym 2-(2-Amino-4-thiazolyl)-(Z)-2-methoxyiminoacetic Acid-d3, Thiazoximic Acid-d3, syn-2-(2-Aminothiazol-4-yl)-2-methoxyiminoacetic Acid-d3, syn-Thiazoximic Acid-d3
InChI formel InChI=1S/C6H7N3O3S/c1-12-9-4(5(10)11)3-2-13-6(7)8-3/h2H,1H3,(H2,7,8)(H,10,11)/b9-4-
SMIL [2H]C([2H])([2H])O\N=C(/C(=O)O)\c1csc(N)n1
IUPAC navn (2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-methoxyiminoacetic acid
Molekylvægt (g/mol) 204.22
Formel vægt 204.0396