Få mere at vide
(-)-2-Amino-6-propionamido-d3-tetrahydrobenzothiazole, TRC
Artikelnummer. 30319432
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30319432 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30319432 Leverandør Toronto Research Chemicals Leverandørnr. TRCA62815225MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (-)-2-Amino-6-propionamido-d3-tetrahydrobenzothiazole
CAS 1217644-20-6
Molekylær formel C10H12D3N3OS
Mængde 25 mg
Synonym N-(2-Amino-4,5,6,7-tetrahydro-benzothiazol-6-yl)propionamide-d3
InChI formel InChI=1S/C10H15N3OS/c1-2-9(14)12-6-3-4-7-8(5-6)15-10(11)13-7/h6H,2-5H2,1H3,(H2,11,13)(H,12,14)/t6-/m0/s1/i1D3
SMIL [2H]C([2H])([2H])CC(=O)N[C@H]1CCc2nc(N)sc2C1
IUPAC navn N-[(6S)-2-amino-4,5,6,7-tetrahydro-1,3-benzothiazol-6-yl]-3,3,3-trideuteriopropanamide
Molekylvægt (g/mol) 228.33
Formel vægt 228.1124