Få mere at vide
2-Amino-6-methyldipyrido[1,2-a:3',2'-d]imidazole Hydrochloride Hydrate, TRC
Artikelnummer. 30136463
Skift visning
Mængde:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30136463 5 mg
30133485 25 mg
17924715 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30136463 Leverandør Toronto Research Chemicals Leverandørnr. TRCA6161005MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 210049-10-8
Molekylær formel C11 H10 N4 . Cl H . H2 O
Tekniske data
Kemisk navn eller materiale 2-Amino-6-methyldipyrido[1,2-a:3',2'-d]imidazole Hydrochloride Hydrate
Mængde 5 mg
Forsendelsestilstand Room Temperature
Formel vægt 252.0778
MDL nummer MFCD00210744
Renhed >95% (HPLC)
Molekylvægt (g/mol) 252.7, 252.70
InChI nøgle WSBNFZWVPJAQMQ-UHFFFAOYSA-N
SMIL O.Cl.CC1=CC=CN2C1=NC1=CC=C(N)N=C21
IUPAC navn 10-methyl-1,3,8-triazatricyclo[7.4.0.0²,⁷]trideca-2,4,6,8,10,12-hexaen-4-amine hydrate hydrochloride