missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-6-methoxybenzothiazole, 98%, Thermo Scientific Chemicals

Catalog Number p-56855
Click to view available options
Quantity:
5 g
50 g
250 g
Packaging:
Glass bottle
This item is not returnable. View return policy
1747-60-0
C8H8N2OS
180.23
MFCD00005787
KZHGPDSVHSDCMX-UHFFFAOYSA-N
2-amino-6-methoxybenzothiazole, 6-methoxybenzo d thiazol-2-amine, 2-benzothiazolamine, 6-methoxy, 6-methoxy-2-aminobenzothiazole, 2-amino-6-methoxy benzothiazole, benzothiazole, 2-amino-6-methoxy, 6-methoxy-2-benzothiazolamine, 6-methoxybenzothiazol-2-ylamine, unii-lat2842yvx, ccris 1393
15630
6-methoxy-1,3-benzothiazol-2-amine
COC1=CC2=C(C=C1)N=C(S2)N
This item is not returnable. View return policy
1747-60-0
C8H8N2OS
180.23
MFCD00005787
KZHGPDSVHSDCMX-UHFFFAOYSA-N
2-amino-6-methoxybenzothiazole, 6-methoxybenzo d thiazol-2-amine, 2-benzothiazolamine, 6-methoxy, 6-methoxy-2-aminobenzothiazole, 2-amino-6-methoxy benzothiazole, benzothiazole, 2-amino-6-methoxy, 6-methoxy-2-benzothiazolamine, 6-methoxybenzothiazol-2-ylamine, unii-lat2842yvx, ccris 1393
15630
6-methoxy-1,3-benzothiazol-2-amine
COC1=CC2=C(C=C1)N=C(S2)N

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 162°C to 169°C
Color Beige to White or Gray-Lilac
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 5 g
Beilstein 27, II, 334
Formula Weight 180.23
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-6-methoxybenzothiazole, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 2-Amino-6-methoxybenzothiazole
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'