missing translation for 'onlineSavingsMsg'
Få mere at vide

2-Amino-6-iodopurine, 97%, Thermo Scientific™

Artikelnummer. 15428137
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
1 g
5 g
Pakningsstørrelse:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
15428137 1 g 1g
15438137 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 15428137 Leverandør Thermo Scientific Alfa Aesar Leverandørnr. H51672.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter

Denne vare kan ikke returneres. Se returpolicy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 19690-23-4
Molekylær formel C5H4IN5
Molekylvægt (g/mol) 261.03
MDL nummer MFCD03840496,MFCD08706375
InChI nøgle CQYPNVKLVHHOSJ-UHFFFAOYSA-N
Synonym 2-amino-6-iodopurine, 6-iodo-9h-purin-2-amine, 2-amino-6-iodo purine, 2-amino-6-iodo-7h-purine, pubchem9213, 6-iodo-2-aminopurine, 2-amino-6-iodo-purine, 6-iodopurine-2-ylamine, acmc-1caa5, amtns010
PubChem CID 6101575
IUPAC navn 6-iodo-7H-purin-2-amine
SMIL NC1=NC(I)=C2NC=NC2=N1

Tekniske data

Kemisk navn eller materiale 2-Amino-6-iodopurine
Mængde 1 g
Følsomhed Light sensitive
Formel vægt 261.02
Procent renhed 97%
Produkttitel
Vælg et problem

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.