missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Amino-6-fluorobenzonitrile, 99%

Brand:  Alfa Aesar™ A12456.06

Additional Details : CAS Number : 77326-36-4 Weight : 0.00500kg Transport : UN number : 3439 Chem class : 6.1 Pack group : III

Quantity 5g

 View more versions of this product

Product Code. 11499683

  • 535.00 DKK / 5g

In stock, ready to ship: 5

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

77326-36-4
136.129
IQUNZGOZUJITBJ-UHFFFAOYSA-N
522659
C1=CC(=C(C(=C1)F)C#N)N
C7H5FN2
MFCD00015477
6-fluoroanthranilonitrile, 2-amino-6-fluorobenzo-nitrile, 2-fluoro-6-aminobenzonitrile, 6-fluoro-2-aminobenzonitrile, 2-amino-6-fluoro-benzonitrile, benzonitrile, 2-amino-6-fluoro, 2-amino-6-fluorobenzenecarbonitrile, pubchem4644, acmc-1bhvi, 2-cyano-3-fluoroaniline
2-amino-6-fluorobenzonitrile

Specifications

2-Amino-6-fluorobenzonitrile
5g
C7H5FN2
UN3439
6-fluoroanthranilonitrile, 2-amino-6-fluorobenzo-nitrile, 2-fluoro-6-aminobenzonitrile, 6-fluoro-2-aminobenzonitrile, 2-amino-6-fluoro-benzonitrile, benzonitrile, 2-amino-6-fluoro, 2-amino-6-fluorobenzenecarbonitrile, pubchem4644, acmc-1bhvi, 2-cyano-3-fluoroaniline
C1=CC(=C(C(=C1)F)C#N)N
136.129
136.13
125°C to 129°C
77326-36-4
MFCD00015477
3588945
IQUNZGOZUJITBJ-UHFFFAOYSA-N
2-amino-6-fluorobenzonitrile
522659
99%
Videos
SDS Documents