Learn More
2-Amino-6-ethylpyridine, 97%, Thermo Scientific Chemicals
Catalog Number p-7024456
Click to view available options
Quantity:
2.5 g
10 g
50 g
21717-29-3
C7H10N2
122.171
MFCD00218265
JXKAUUVMXZIJNZ-UHFFFAOYSA-N
2-amino-6-ethylpyridine, 6-ethyl-2-pyridinamine, 2-pyridinamine, 6-ethyl, 6-ethyl-2-pyridylamine, acmc-1clku, 2-pyridinamine,6-ethyl, 2-amino-6-ethyl-pyridine, 6-ethyl-pyridin-2-ylamine
89021
6-ethylpyridin-2-amine
CCC1=NC(=CC=C1)N
21717-29-3
C7H10N2
122.171
MFCD00218265
JXKAUUVMXZIJNZ-UHFFFAOYSA-N
2-amino-6-ethylpyridine, 6-ethyl-2-pyridinamine, 2-pyridinamine, 6-ethyl, 6-ethyl-2-pyridylamine, acmc-1clku, 2-pyridinamine,6-ethyl, 2-amino-6-ethyl-pyridine, 6-ethyl-pyridin-2-ylamine
89021
6-ethylpyridin-2-amine
CCC1=NC(=CC=C1)N
The action of iodine on 2-amino-6-ethylpyridine in an alkaline medium gives 2-amino-5-iodo-6-ethylpyridine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
The action of iodine on 2-amino-6-ethylpyridine in an alkaline medium gives 2-amino-5-iodo-6-ethylpyridine.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Odor Unpleasant
Quantity 2.5 g
UN Number UN2810
Formula Weight 122.17
Percent Purity 97%
Chemical Name or Material 2-Amino-6-ethylpyridine