missing translation for 'onlineSavingsMsg'
Få mere at vide

2-Amino-6-chloro-9-(2,3,5-tri-O-acetyl-Beta-D-ribofuranosyl)purine, TRC

Artikelnummer. 30310463
Change view
Click to view available options
Mængde:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30310463 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30310463 Supplier Toronto Research Chemicals Supplier No. TRCA6035155G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 2-Amino-6-chloro-9-(2,3,5-tri-O-acetyl-beta-D-ribofuranosyl)purine
CAS 16321-99-6
Molekylær formel C16 H18 Cl N5 O7
Mængde 5 g
InChI formel InChI=1S/C16H18ClN5O7/c1-6(23)26-4-9-11(27-7(2)24)12(28-8(3)25)15(29-9)22-5-19-10-13(17)20-16(18)21-14(10)22/h5,9,11-12,15H,4H2,1-3H3,(H2,18,20,21)/t9-,11-,12-,15-/m1/s1
SMIL CC(=O)OC[C@H]1O[C@H]([C@H](OC(=O)C)[C@@H]1OC(=O)C)n2cnc3c(Cl)nc(N)nc23
IUPAC navn [(2R,3R,4R,5R)-3,4-diacetyloxy-5-(2-amino-6-chloropurin-9-yl)oxolan-2-yl]methyl acetate
Molekylvægt (g/mol) 427.8
Formel vægt 427.0895
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.