missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Amino-6-bromopyridine, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
Employed in an efficient one-pot synthesis of 7-azaindoles. Used in the synthesis of anti-HIV agents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Employed in an efficient one-pot synthesis of 7-azaindoles. Used in the synthesis of anti-HIV agents.
Solubility
Slightly soluble in water.
Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Employed in an efficient one-pot synthesis of 7-azaindoles. Used in the synthesis of anti-HIV agents.
Solubility
Slightly soluble in water.
Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Melting Point | 88°C to 91°C |
Quantity | 5 g |
Solubility Information | Slightly soluble in water. |
Formula Weight | 173.01 |
Percent Purity | 98% |
Chemical Name or Material | 2-Amino-6-bromopyridine |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'