missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-tert-butyl-1,3,4-thiadiazole, 97%, Thermo Scientific Chemicals

Catalog Number p-7044386
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
39222-73-6
C6H11N3S
157.24
MFCD00040403
ICXDPEFCLDSXLI-UHFFFAOYSA-N
2-amino-5-tert-butyl-1,3,4-thiadiazole, buthidazolamine, 5-tert-butyl-1,3,4-thiadiazol-2-amine, 1,3,4-thiadiazol-2-amine, 5-1,1-dimethylethyl, unii-x53j1t94sn, 5-tert-butyl-1,3,4 thiadiazol-2-ylamine, 5-tert-butyl-1,3,4-thiadiazole-2-ylamine, acmc-20aidm, enamine_005678, 5-t-butyl-2-amino1,3,4-thiadiazole
170170
5-tert-butyl-1,3,4-thiadiazol-2-amine
CC(C)(C)C1=NN=C(N)S1
This item is not returnable. View return policy
39222-73-6
C6H11N3S
157.24
MFCD00040403
ICXDPEFCLDSXLI-UHFFFAOYSA-N
2-amino-5-tert-butyl-1,3,4-thiadiazole, buthidazolamine, 5-tert-butyl-1,3,4-thiadiazol-2-amine, 1,3,4-thiadiazol-2-amine, 5-1,1-dimethylethyl, unii-x53j1t94sn, 5-tert-butyl-1,3,4 thiadiazol-2-ylamine, 5-tert-butyl-1,3,4-thiadiazole-2-ylamine, acmc-20aidm, enamine_005678, 5-t-butyl-2-amino1,3,4-thiadiazole
170170
5-tert-butyl-1,3,4-thiadiazol-2-amine
CC(C)(C)C1=NN=C(N)S1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 181°C to 185°C
Quantity 1 g
Beilstein 119853
Formula Weight 157.24
Percent Purity 97%
Chemical Name or Material 2-Amino-5-tert-butyl-1,3,4-thiadiazole

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'