Få mere at vide
(2-Amino-5-nitrophenyl)phenyl-methanone (2-Amino-5-nitrobenzophenone), Mikromol™
Artikelnummer. 30050243
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30050243 100 mg
30050006 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30050243 Leverandør Mikromol Leverandørnr. MM0078.01
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
(2-Amino-5-nitrophenyl)phenyl-methanone (2-Amino-5-nitrobenzophenone)
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Amino-5-nitrobenzophenone
CAS 1775-95-7
Analyt- eller komponentnavne 2-Amino-5-nitrobenzophenone
Forsendelsestilstand Room Temperature
Molekylær formel C13 H10 N2 O3
Certificeringer/compliance ISO 17025
Synonym (2-Amino-5-nitrophenyl)phenyl-methanone, Nitrazepam Imp. B (EP), Nitrazepam Impurity B, 5-Nitro-2-aminobenzophenone, Ro 7-19990
InChI formel InChI=1S/C13H10N2O3/c14-12-7-6-10(15(17)18)8-11(12)13(16)9-4-2-1-3-5-9/h1-8H,14H2
SMIL Nc1ccc(cc1C(=O)c2ccccc2)[N+](=O)[O-]
IUPAC navn (2-amino-5-nitrophenyl)-phenylmethanone
Molekylvægt (g/mol) 242.23
Mængde 100 mg
Formel vægt 242.0691 g/mol