Få mere at vide
2-Amino-5-nitrobenzophenone, LoGiCal
Artikelnummer. 30463436
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30463436 10 mg
1 options
Artikelnummer. 30463436 Leverandør LoGiCal Leverandørnr. LGCFOR0078.01
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Amino-5-nitrobenzophenone
CAS 1775-95-7
Analyt- eller komponentnavne 2-Amino-5-nitrobenzophenone
Forsendelsestilstand Room Temperature
Molekylær formel C13 H10 N2 O3
Certificeringer/compliance ISO 9001
Synonym (2-Amino-5-nitrophenyl)phenyl-methanone, Nitrazepam Imp. B (EP), Nitrazepam Impurity B, 5-Nitro-2-aminobenzophenone, Ro 7-19990
InChI formel InChI=1S/C13H10N2O3/c14-12-7-6-10(15(17)18)8-11(12)13(16)9-4-2-1-3-5-9/h1-8H,14H2
Løsningstype Neat
SMIL Nc1ccc(cc1C(=O)c2ccccc2)[N+](=O)[O-]
IUPAC navn (2-amino-5-nitrophenyl)-phenylmethanone
Industry Type Clinical Testing
Molekylvægt (g/mol) 242.23
Mængde 10 mg
Formel vægt 242.0691 g/mol