Få mere at vide
2-Amino-5-methylhexanoic Acid, TRC
Artikelnummer. 30240798
Skift visning
Mængde:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30240798 500 mg
30240765 100 mg
30240510 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30240798 Leverandør Toronto Research Chemicals Leverandørnr. TRCA192141500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 3558-32-5
Molekylær formel C7 H15 N O2
Molekylvægt (g/mol) 145.2
Synonym 5-Methylnorleucine, (±)-2-Amino-5-methylhexanoic Acid, 2-Amino-5-methylhexanoic Acid, 5-Methyl-DL-norleucine, DL-2-Amino-5-methylhexanoic Acid, Homoleucine, NSC 20145
IUPAC navn 2-amino-5-methylhexanoic acid
SMIL CC(C)CCC(N)C(=O)O
Tekniske data
Kemisk navn eller materiale 2-Amino-5-methylhexanoic Acid
InChI formel InChI=1S/C7H15NO2/c1-5(2)3-4-6(8)7(9)10/h5-6H,3-4,8H2,1-2H3,(H,9,10)
Mængde 500 mg
Formel vægt 145.1103