Få mere at vide
2-Amino-5-methoxybenzimidazole, TRC
Artikelnummer. 30133419
Skift visning
Mængde:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30133419 1 g
30010281 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30133419 Leverandør Toronto Research Chemicals Leverandørnr. TRCA6119201G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 6232-91-3
Molekylær formel C8H9N3O
Molekylvægt (g/mol) 163.18
Synonym 5-Methoxy-1H-benzimidazol-2-amine, 5-Methoxy-2-aminobenzimidazole, 5-Methoxybenzimidazole-2-amine, 6-Methoxy-1H-benzimidazol-2-amine
IUPAC navn 6-methoxy-1H-benzimidazol-2-amine
SMIL COC1=CC2=C(C=C1)N=C(N)N2
Tekniske data
Kemisk navn eller materiale 2-Amino-5-methoxybenzimidazole
Mængde 1 g
InChI formel InChI=1S/C8H9N3O/c1-12-5-2-3-6-7(4-5)11-8(9)10-6/h2-4H,1H3,(H3,9,10,11)
Formel vægt 163.0746