missing translation for 'onlineSavingsMsg'
Få mere at vide

2-Amino-5-iodo-3-methylbenzoic Acid, TRC

Artikelnummer. 30309329
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30309329 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30309329 Leverandør Toronto Research Chemicals Leverandørnr. TRCA6041255G

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale 2-Amino-5-iodo-3-methylbenzoic Acid
CAS 108857-24-5
Molekylær formel C8H8INO2
Mængde 5 g
InChI formel InChI=1S/C8H8INO2/c1-4-2-5(9)3-6(7(4)10)8(11)12/h2-3H,10H2,1H3,(H,11,12)
SMIL Cc1cc(I)cc(C(=O)O)c1N
IUPAC navn 2-amino-5-iodo-3-methylbenzoic acid
Molekylvægt (g/mol) 277.06
Formel vægt 276.96
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.